Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP8B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004391.2 | NP_004382.2 | 1 | 2174 | ||
GenBank mRNA | AF090318.1 | - | 1 | 1733 |