Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004398.3 | NP_004389.2 | 6 | 897 | ||
GenBank mRNA | AB001343.1 | - | 2 | 196 | ||
AB040537.1 | - | 6 | 833 | |||
AB209884.1 | - | 6 | 800 | |||
AK296752.1 | - | 4 | 571 | |||
AK315216.1 | - | 6 | 828 | |||
BC049217.1 | - | 6 | 793 | |||
BC091521.1 | - | 6 | 845 | 30388370 | ||
BC093654.1 | - | 6 | 801 | 7939499 | ||
BC093656.1 | - | 6 | 801 | 7939501 | ||
U28042.1 | - | 6 | 833 |