Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127453.1 | NP_001120925.1 | 10 | 1648 | ||
NM_001127454.1 | NP_001120926.1 | 9 | 1663 | |||
NM_004403.2 | NP_004394.1 | 10 | 1893 | |||
XM_017011802.1 | XP_016867291.1 | 9 | 1419 | |||
GenBank mRNA | AF007790.2 | - | 9 | 1548 | ||
AF073308.1 | - | 10 | 1617 | |||
AF075171.1 | - | 2 | 1302 | |||
AF131765.1 | - | 2 | 1304 | |||
AK094714.1 | - | 12 | 2221 | |||
BC099911.1 | - | 10 | 1609 | 5287362 | ||
BC125065.1 | - | 10 | 1615 | 40117291 | ||
BC125066.1 | - | 9 | 1385 | |||
BX647389.1 | - | 2 | 3580 |