Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285389.2 | NP_001272318.1 | 4 | 508 | ||
NM_001285391.2 | NP_001272320.2 | 4 | 831 | |||
NM_001285392.2 | NP_001272321.1 | 4 | 625 | |||
NM_001285393.2 | NP_001272322.1 | 4 | 562 | |||
NM_004414.6 | NP_004405.3 | 4 | 834 | |||
NM_203417.2 | NP_981962.1 | 4 | 620 | |||
NM_203418.2 | NP_981963.1 | 4 | 799 | |||
XM_017028283.1 | XP_016883772.1 | 5 | 1160 | |||
GenBank mRNA | AF400429.1 | - | 4 | 492 | ||
AK055845.1 | - | 3 | 1031 | |||
AK092184.1 | - | 4 | 755 | |||
AK131569.1 | - | 4 | 752 | |||
AK299444.1 | - | 4 | 549 | |||
AL556803.3 | - | 4 | 834 | |||
AY325903.1 | - | 4 | 826 | |||
BC002864.2 | - | 4 | 594 | 3944959 | ||
BT007363.1 | - | 4 | 538 | |||
BX419260.2 | - | 4 | 752 | |||
CR456878.1 | - | 3 | 460 | |||
CR542065.1 | - | 4 | 538 | |||
U28833.2 | - | 4 | 508 | |||
U85265.2 | - | 4 | 619 | |||
U85266.2 | - | 4 | 562 | |||
U85267.2 | - | 4 | 681 |