Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004423.3 | NP_004414.3 | 8 | 1034 | ||
XM_005247172.2 | XP_005247229.1 | 8 | 1219 | |||
XM_011512513.2 | XP_011510815.1 | 5 | 822 | |||
GenBank mRNA | AF006013.1 | - | 8 | 905 | ||
AK093469.1 | - | 8 | 1026 | |||
AK304686.1 | - | 9 | 986 | |||
AL110278.1 | - | 4 | 676 | |||
BC018289.1 | - | 6 | 570 | |||
BC032459.1 | - | 8 | 962 | 5200610 | ||
D86963.1 | - | 8 | 1034 | |||
U48252.1 | - | 1 | 139 | |||
U49262.1 | - | 8 | 1015 | |||
U75651.1 | - | 8 | 1287 |