Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004428.2 | NP_004419.2 | 5 | 1412 | ||
NM_182685.1 | NP_872626.1 | 4 | 1346 | |||
GenBank mRNA | AK057845.1 | - | 5 | 1825 | ||
AK289589.1 | - | 4 | 1343 | |||
AL833641.1 | - | 4 | 1344 | |||
BC032698.1 | - | 5 | 1405 | 5402648 | ||
BC095432.1 | - | 5 | 1394 | 30376031 | ||
M57730.1 | - | 5 | 1380 |