Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004429.4 | NP_004420.1 | 5 | 2824 | ||
GenBank mRNA | BC016649.1 | - | 5 | 2822 | 3867060 | |
BC052979.1 | - | 5 | 2845 | 6164440 | ||
U09303.1 | - | 5 | 2744 |