Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 5 | 535 | ||
NM_004434.2 | NP_004425.2 | 4 | 478 | |||
XM_005267397.1 | XP_005267454.1 | 4 | 548 | |||
XM_005267398.3 | XP_005267455.1 | 4 | 481 | |||
XM_005267399.4 | XP_005267456.2 | 4 | 520 | |||
XM_005267400.4 | XP_005267457.1 | 4 | 504 | |||
XM_011536540.2 | XP_011534842.1 | 4 | 551 | |||
XM_011536541.1 | XP_011534843.1 | 4 | 561 | |||
XM_011536542.2 | XP_011534844.1 | 4 | 520 | |||
XM_017021074.1 | XP_016876563.1 | 4 | 548 | |||
GenBank mRNA | AK126980.1 | - | 6 | 491 | ||
AK299812.1 | - | 5 | 814 | |||
AK307925.1 | - | 4 | 471 | |||
BC032541.1 | - | 4 | 454 | 5533599 | ||
BC033043.1 | - | 4 | 478 | 4821202 | ||
BX247979.1 | - | 5 | 547 | |||
BX420178.2 | - | 5 | 546 | |||
U97018.1 | - | 4 | 476 |