Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KRIT1 SIRNA Assays ›
Gene Symbol:
KRIT1
Gene Name:
KRIT1, ankyrin repeat containing
Gene Aliases:
CAM, CCM1
Chromosome Location:
Chr.7: 92198969 - 92246128 on Build GRCh38
UniGene ID:
Hs.531987
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146529
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KRIT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013406.1
NP_001013424.1
4
604
NM_004912.3
NP_004903.2
5
1099
NM_194454.1
NP_919436.1
5
752
NM_194455.1
NP_919437.1
5
1226
NM_194456.1
NP_919438.1
6
991
XM_005250660.3
XP_005250717.1
5
719
XM_005250662.3
XP_005250719.1
5
698
XM_005250665.3
XP_005250722.1
5
626
XM_005250666.3
XP_005250723.1
6
802
XM_005250667.2
XP_005250724.1
4
510
XM_005250668.3
XP_005250725.1
4
572
XM_005250669.3
XP_005250726.1
4
550
XM_006716161.3
XP_006716224.1
5
850
XM_006716162.3
XP_006716225.1
5
667
XM_006716163.3
XP_006716226.1
4
517
XM_011516651.2
XP_011514953.1
6
833
XM_011516653.2
XP_011514955.1
4
587
XM_011516654.2
XP_011514956.1
6
851
XM_011516655.2
XP_011514957.1
4
505
XM_011516656.2
XP_011514958.1
4
474
XM_011516657.2
XP_011514959.1
4
394
XM_011516658.2
XP_011514960.1
4
406
XM_011516659.2
XP_011514961.1
4
428
XM_011516660.2
XP_011514962.1
3
310
XM_011516661.2
XP_011514963.1
3
388
XM_017012755.1
XP_016868244.1
4
477
XM_017012756.1
XP_016868245.1
4
439
XM_017012757.1
XP_016868246.1
4
471
XM_017012758.1
XP_016868247.1
5
1077
XM_017012759.1
XP_016868248.1
4
929
GenBank mRNA
AF296765.2
-
5
996
AF310133.1
-
5
691
AF388384.1
-
5
1226
AJ294850.1
-
6
950
AK055305.1
-
4
604
AK056537.1
-
4
534
AY380057.1
-
5
691
AY380058.1
-
5
691
AY380059.1
-
5
691
AY380060.1
-
5
691
AY993944.1
-
2
207
BC094684.1
-
2
217
6174749
BX648659.1
-
1
72
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of endothelial cell proliferation
small GTPase mediated signal transduction
negative regulation of endothelial cell migration
negative regulation of angiogenesis
positive regulation of protein binding
cell redox homeostasis
regulation of catalytic activity
regulation of establishment of cell polarity
negative regulation of endothelial cell apoptotic process
protein binding
phosphatidylinositol-4,5-bisphosphate binding
microtubule binding
GTPase regulator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-xxl4s:80/100.66.77.167:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline