Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004947.4 | NP_004938.1 | 8, 9 | 622 | ||
XM_005264914.3 | XP_005264971.1 | 8, 9 | 894 | |||
XM_005264915.3 | XP_005264972.1 | 8, 9 | 894 | |||
XM_005264916.3 | XP_005264973.1 | 8, 9 | 672 | |||
XM_005264917.3 | XP_005264974.1 | 8, 9 | 894 | |||
XM_005264918.3 | XP_005264975.1 | 8, 9 | 671 | |||
XM_006713008.3 | XP_006713071.1 | 8, 9 | 894 | |||
XM_006713009.3 | XP_006713072.1 | 8, 9 | 894 | |||
XM_006713010.3 | XP_006713073.1 | 8, 9 | 657 | |||
XM_017005825.1 | XP_016861314.1 | 8, 9 | 673 | |||
XM_017005826.1 | XP_016861315.1 | 8, 9 | 663 | |||
GenBank mRNA | AB002297.1 | - | 3, 4 | 230 | ||
AY145303.1 | - | 8, 9 | 599 | |||
AY254099.1 | - | 8, 9 | 622 |