Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004947.4 | NP_004938.1 | 30 | 3181 | ||
XM_005264914.3 | XP_005264971.1 | 30 | 3453 | |||
XM_005264915.3 | XP_005264972.1 | 30 | 3453 | |||
XM_005264916.3 | XP_005264973.1 | 30 | 3231 | |||
XM_005264917.3 | XP_005264974.1 | 30 | 3411 | |||
XM_005264918.3 | XP_005264975.1 | 29 | 3083 | |||
XM_006713008.3 | XP_006713071.1 | 30 | 3453 | |||
XM_006713009.3 | XP_006713072.1 | 30 | 3453 | |||
XM_006713010.3 | XP_006713073.1 | 30 | 3216 | |||
XM_011533441.2 | XP_011531743.1 | 19 | 4062 | |||
XM_011533443.2 | XP_011531745.1 | 6 | 1743 | |||
XM_011533444.2 | XP_011531746.1 | 6 | 1667 | |||
XM_011533445.2 | XP_011531747.1 | 6 | 1688 | |||
XM_017005825.1 | XP_016861314.1 | 30 | 3232 | |||
XM_017005826.1 | XP_016861315.1 | 30 | 3222 | |||
XM_017005827.1 | XP_016861316.1 | 21 | 2373 | |||
XM_017005828.1 | XP_016861317.1 | 5 | 1098 | |||
GenBank mRNA | AB002297.1 | - | 25 | 2789 | ||
AY145303.1 | - | 30 | 3158 | |||
AY254099.1 | - | 30 | 3181 |