Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164617.1 | NP_001158089.1 | 2, 3 | 544 | ||
NM_001164619.1 | NP_001158091.1 | 1, 2 | 382 | |||
NM_004484.3 | NP_004475.1 | 2, 3 | 544 | |||
XM_017029413.1 | XP_016884902.1 | 2, 3 | 515 | |||
GenBank mRNA | AK222761.1 | - | 2, 3 | 492 | ||
AK222766.1 | - | 2, 3 | 492 | |||
AK310196.1 | - | 1, 2 | 211 | |||
AK310689.1 | - | 3, 4 | 563 | |||
BC035972.1 | - | 2, 3 | 492 | 4603748 | ||
DQ349136.1 | - | 1, 2 | 185 | |||
DQ349137.1 | - | 2, 3 | 347 | |||
DQ349138.1 | - | 2, 3 | 347 | |||
JQ943686.1 | - | 2, 3 | 347 | |||
L47125.1 | - | 2, 3 | 498 | |||
L47176.1 | - | 2, 3 | 493 | |||
U50410.1 | - | 2, 3 | 491 | |||
Z37987.1 | - | 2, 3 | 455 |