Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004486.4 | NP_004477.3 | 2, 3 | 225 | ||
XM_005251932.1 | XP_005251989.1 | 2, 3 | 236 | |||
GenBank mRNA | AF248953.1 | - | 2, 3 | 176 | ||
BC069268.1 | - | 2, 3 | 225 | 6340972 |