Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFA2T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005187.5 | NP_005178.4 | 12 | 4276 | ||
NM_175931.2 | NP_787127.1 | 11 | 3942 | |||
XM_005256323.4 | XP_005256380.1 | 11 | 4254 | |||
GenBank mRNA | AB010419.1 | - | 12 | 4145 | ||
AB010420.1 | - | 11 | 3942 |