Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005191.3 | NP_005182.1 | 6 | 1248 | ||
XM_011513327.2 | XP_011511629.1 | 6 | 3314 | |||
GenBank mRNA | AK292129.1 | - | 6 | 1232 | ||
AY197777.1 | - | 4 | 757 | |||
AY197778.1 | - | 3 | 475 | |||
BC042665.1 | - | 6 | 1216 | 5181343 | ||
M27533.1 | - | 6 | 1170 |