Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085471.1 | NP_001078940.1 | 2 | 259 | ||
NM_005197.3 | NP_005188.2 | 2 | 376 | |||
GenBank mRNA | AI240081.1 | - | 2 | 296 | ||
AK225967.1 | - | 3 | 349 | |||
AK308053.1 | - | 2 | 258 | |||
BC007506.2 | - | 2 | 270 | 3028787 | ||
BP213901.1 | - | 3 | 368 | |||
CB989352.1 | - | 2 | 403 | |||
U68723.1 | - | 2 | 254 |