Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085471.1 | NP_001078940.1 | 7 | 1664 | ||
NM_005197.3 | NP_005188.2 | 6 | 1715 | |||
GenBank mRNA | AK225967.1 | - | 8 | 1449 | ||
BC007506.2 | - | 6 | 1609 | 3028787 | ||
U68723.1 | - | 7 | 1659 |