Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288705.1 | NP_001275634.1 | 6 | 1265 | ||
NM_005211.3 | NP_005202.2 | 7 | 1344 | |||
NR_109969.1 | 6 | 1265 | ||||
GenBank mRNA | AB208819.1 | - | 6 | 1215 | ||
AK304491.1 | - | 6 | 1046 | |||
BC129939.1 | - | 6 | 1185 | |||
BX648599.1 | - | 5 | 1135 | |||
CD620053.1 | - | 2 | 337 | |||
X03663.1 | - | 7 | 1352 |