Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288705.1 | NP_001275634.1 | 21 | 3844 | ||
NM_005211.3 | NP_005202.2 | 22 | 3923 | |||
NR_109969.1 | 20 | 3681 | ||||
GenBank mRNA | AB208819.1 | - | 20 | 3632 | ||
BC047521.1 | - | 21 | 3827 | 4819527 | ||
BX648599.1 | - | 20 | 3715 | |||
X03663.1 | - | 22 | 3931 |