Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTLA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037631.2 | NP_001032720.1 | 3 | 867 | ||
NM_005214.4 | NP_005205.2 | 4 | 977 | |||
GenBank mRNA | AF414120.1 | - | 4 | 977 | ||
BC069566.1 | - | 4 | 917 | 7262243 |