Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004490.2 | NP_004481.2 | 1, 2 | 740 | ||
XM_011511022.2 | XP_011509324.1 | 1, 2 | 372 | |||
GenBank mRNA | AK307744.1 | - | 1, 2 | 252 | ||
AK313153.1 | - | 1, 2 | 369 | |||
BC053559.1 | - | 1, 2 | 210 | 6162863 | ||
L76687.1 | - | 1, 2 | 739 |