Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005215.3 | NP_005206.2 | 2 | 730 | ||
XM_011525843.1 | XP_011524145.1 | 2 | 730 | |||
XM_017025568.1 | XP_016881057.1 | 2 | 730 | |||
XM_017025569.1 | XP_016881058.1 | 2 | 730 | |||
GenBank mRNA | M32292.1 | - | 2 | 120 | ||
X76132.1 | - | 2 | 114 |