Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079812.2 | NP_001073280.1 | 1 | 245 | ||
NM_001314007.1 | NP_001300936.1 | 1 | 245 | |||
NM_005219.4 | NP_005210.3 | 1 | 245 | |||
GenBank mRNA | AB209482.1 | - | 1 | 186 | ||
AF051782.1 | - | 1 | 104 | |||
AK304291.1 | - | 1 | 191 | |||
AY363395.1 | - | 1 | 104 | |||
BC117257.1 | - | 1 | 122 | 40125808 | ||
BC143413.1 | - | 1 | 122 | 9051921 | ||
BC143414.1 | - | 1 | 122 | |||
BQ945734.1 | - | 1 | 179 | |||
DA593734.1 | - | 1 | 245 | |||
DQ067453.1 | - | 1 | 119 |