Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079812.2 | NP_001073280.1 | 5 | 711 | ||
NM_001314007.1 | NP_001300936.1 | 6 | 738 | |||
NM_005219.4 | NP_005210.3 | 6 | 738 | |||
XM_011537572.2 | XP_011535874.1 | 5 | 679 | |||
XM_011537573.2 | XP_011535875.1 | 6 | 546 | |||
GenBank mRNA | AB209482.1 | - | 6 | 679 | ||
AF051782.1 | - | 5 | 570 | |||
AK307822.1 | - | 2 | 170 | |||
AY363395.1 | - | 6 | 597 | |||
BC117257.1 | - | 5 | 588 | 40125808 | ||
BC143413.1 | - | 6 | 615 | 9051921 | ||
BC143414.1 | - | 6 | 615 |