Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005232.4 | NP_005223.4 | 6 | 1342 | ||
GenBank mRNA | AK290351.1 | - | 6 | 1306 | ||
AK313263.1 | - | 6 | 1315 | |||
BC130291.1 | - | 6 | 1326 | 40146322 | ||
BC144156.1 | - | 6 | 1326 | |||
CD653381.1 | - | 1 | 231 | |||
EU826604.1 | - | 6 | 1255 | |||
EU826605.1 | - | 6 | 1255 | |||
M18391.1 | - | 6 | 1348 | |||
Z27409.1 | - | 2 | 400 |