Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005233.5 | NP_005224.2 | 3, 4 | 1047 | ||
NM_182644.2 | NP_872585.1 | 3, 4 | 1047 | |||
XM_005264715.2 | XP_005264772.1 | 3, 4 | 972 | |||
XM_005264716.2 | XP_005264773.1 | 3, 4 | 1042 | |||
GenBank mRNA | AF213459.1 | - | 3, 4 | 1047 | ||
AF213460.1 | - | 3, 4 | 909 | |||
AK291411.1 | - | 3, 4 | 880 | |||
BC063282.1 | - | 3, 4 | 951 | 30347891 | ||
M83941.1 | - | 3, 4 | 922 |