Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308028.1 | NP_001294957.1 | 4 | 514 | ||
NM_005246.3 | NP_005237.2 | 6 | 900 | |||
XM_011543266.1 | XP_011541568.1 | 6 | 753 | |||
XM_011543267.1 | XP_011541569.1 | 6 | 2637 | |||
XM_011543269.1 | XP_011541571.1 | 5 | 2491 | |||
XM_011543270.2 | XP_011541572.1 | 5 | 668 | |||
XM_011543271.2 | XP_011541573.1 | 6 | 753 | |||
XM_011543272.1 | XP_011541574.1 | 6 | 753 | |||
XM_011543278.1 | XP_011541580.1 | 6 | 753 | |||
XM_017009229.1 | XP_016864718.1 | 7 | 900 | |||
XM_017009230.1 | XP_016864719.1 | 5 | 668 | |||
XM_017009231.1 | XP_016864720.1 | 5 | 2491 | |||
XM_017009232.1 | XP_016864721.1 | 5 | 541 | |||
XM_017009233.1 | XP_016864722.1 | 4 | 484 | |||
XM_017009234.1 | XP_016864723.1 | 6 | 753 | |||
XM_017009235.1 | XP_016864724.1 | 6 | 753 | |||
GenBank mRNA | AK293376.1 | - | 5 | 754 | ||
AK296874.1 | - | 5 | 475 | |||
AK299855.1 | - | 4 | 479 | |||
AK315234.1 | - | 4 | 588 | |||
BC017060.1 | - | 4 | 582 | |||
J03358.1 | - | 6 | 900 |