Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006135.2 | NP_006126.1 | 10 | 1308 | ||
XM_017002424.1 | XP_016857913.1 | 10 | 1148 | |||
GenBank mRNA | AK223226.1 | - | 10 | 929 | ||
AK289654.1 | - | 10 | 951 | |||
BC000144.1 | - | 10 | 933 | 3047937 | ||
BX648738.1 | - | 10 | 1599 | |||
U56637.1 | - | 10 | 916 |