Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006137.6 | NP_006128.1 | 4 | 1257 | ||
XM_011523608.1 | XP_011521910.1 | 4 | 1224 | |||
XM_017025316.1 | XP_016880805.1 | 4 | 1290 | |||
GenBank mRNA | AK310558.1 | - | 3 | 2001 | ||
AY935534.1 | - | 4 | 1229 | |||
AY935535.1 | - | 4 | 1229 | |||
AY935536.1 | - | 4 | 1229 | |||
AY935537.1 | - | 4 | 1217 | |||
BC009293.2 | - | 4 | 1201 | 4120480 | ||
BC013297.2 | - | 4 | 1201 | 3529600 | ||
X06180.1 | - | 3 | 1634 |