Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FKTN SIRNA Assays ›
Gene Symbol:
FKTN
Gene Name:
fukutin
Gene Aliases:
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
Chromosome Location:
Chr.9: 105558117 - 105655950 on Build GRCh38
UniGene ID:
Hs.55777
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146692
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FKTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079802.1
NP_001073270.1
6
668
NM_001198963.1
NP_001185892.1
6
668
NM_006731.2
NP_006722.2
5
576
XM_006717014.2
XP_006717077.1
5
580
XM_011518368.2
XP_011516670.1
7
816
XM_011518369.2
XP_011516671.1
6
666
XM_011518370.2
XP_011516672.1
7
2352
XM_011518371.2
XP_011516673.1
7
851
XM_011518373.2
XP_011516675.1
7
762
XM_011518374.2
XP_011516676.1
5
580
XM_011518375.2
XP_011516677.1
5
895
XM_011518376.2
XP_011516678.1
5
718
XM_011518378.2
XP_011516680.1
7
816
XM_011518379.2
XP_011516681.1
5
462
XM_011518381.2
XP_011516683.1
6
651
XM_011518387.2
XP_011516689.1
7
816
XM_011518389.2
XP_011516691.1
7
816
XM_011518390.2
XP_011516692.1
3
257
XM_011518391.2
XP_011516693.1
7
816
XM_017014462.1
XP_016869951.1
8
950
XM_017014463.1
XP_016869952.1
5
1039
XM_017014464.1
XP_016869953.1
7
816
XM_017014465.1
XP_016869954.1
6
666
XM_017014466.1
XP_016869955.1
7
816
XM_017014467.1
XP_016869956.1
6
1075
XM_017014468.1
XP_016869957.1
5
958
XM_017014469.1
XP_016869958.1
7
816
XM_017014470.1
XP_016869959.1
7
816
XM_017014471.1
XP_016869960.1
8
914
XM_017014472.1
XP_016869961.1
8
940
XM_017014473.1
XP_016869962.1
3
2818
XM_017014474.1
XP_016869963.1
5
579
XM_017014475.1
XP_016869964.1
5
583
XM_017014476.1
XP_016869965.1
8
934
XM_017014477.1
XP_016869966.1
7
799
XM_017014478.1
XP_016869967.1
9
1001
XM_017014479.1
XP_016869968.1
6
698
GenBank mRNA
AB008226.1
-
5
563
AB609007.1
-
563
AK300840.1
-
6
666
AK304460.1
-
5
574
AL601876.1
-
5
579
BC101808.1
-
4
478
8069314
BC112038.1
-
4
478
8327506
BC117699.1
-
4
484
40073633
BC117700.1
-
6
726
40073634
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nervous system development
muscle organ development
negative regulation of cell proliferation
protein O-linked mannosylation
negative regulation of JNK cascade
regulation of protein glycosylation
transferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0