Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291485.1 | NP_001278414.1 | 5 | 1856 | ||
NM_006890.4 | NP_008821.2 | 5 | 1876 | |||
GenBank mRNA | X98311.1 | - | 5 | 1766 |