Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201478.1 | NP_001188407.1 | 6, 7 | 783 | ||
NM_006893.2 | NP_008824.2 | 8, 9 | 1155 | |||
XM_005277509.3 | XP_005277566.1 | 8, 9 | 1070 | |||
XM_006711196.3 | XP_006711259.1 | 8, 9 | 1026 | |||
XM_011509257.2 | XP_011507559.1 | 8, 9 | 1070 | |||
XM_017000530.1 | XP_016856019.1 | 5, 6 | 590 | |||
XM_017000531.1 | XP_016856020.1 | 6, 7 | 698 | |||
XM_017000532.1 | XP_016856021.1 | 8, 9 | 1026 | |||
GenBank mRNA | AF159586.1 | - | 3, 4 | 246 | ||
AF220417.2 | - | 8, 9 | 1063 | |||
AF262403.1 | - | 6, 7 | 691 | |||
AK001852.1 | - | 8, 9 | 1068 | |||
AK225259.1 | - | 8, 9 | 1049 | |||
AL162001.1 | - | 3, 4 | 361 | |||
BC001585.1 | - | 8, 9 | 1053 | 3463026 | ||
BC039134.1 | - | 8, 9 | 1006 | 4339127 | ||
BC058905.1 | - | 8, 9 | 1020 | 6067337 |