Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320917.1 | NP_001307846.1 | 2 | 3056 | ||
NM_012125.3 | NP_036257.1 | 3 | 2814 | |||
GenBank mRNA | BC041805.1 | - | 2 | 2458 | 5298446 | |
BC068528.1 | - | 2 | 2452 | 30339053 | ||
BX102692.1 | - | 1 | 187 |
Set Membership: |
G-protein coupled receptor |