Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306090.1 | NP_001293019.1 | 8 | 1255 | ||
NM_001306092.1 | NP_001293021.1 | 8 | 1255 | |||
NM_153717.2 | NP_714928.1 | 8 | 1255 | |||
XM_006713865.3 | XP_006713928.1 | 8 | 1252 | |||
XM_006713866.3 | XP_006713929.1 | 8 | 1252 | |||
XM_011513419.2 | XP_011511721.1 | 8 | 1249 | |||
XM_017007883.1 | XP_016863372.1 | 8 | 1249 | |||
GenBank mRNA | AF216184.1 | - | 8 | 1255 | ||
AF216185.1 | - | 8 | 1255 | |||
AF239742.1 | - | 8 | 1071 | |||
AK025394.1 | - | 3 | 384 |