Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020730.2 | NP_065781.1 | 3 | 636 | ||
NM_021120.3 | NP_066943.2 | 9 | 1677 | |||
XM_005262248.3 | XP_005262305.1 | 3 | 233 | |||
XM_006724625.2 | XP_006724688.1 | 9 | 1682 | |||
XM_006724626.2 | XP_006724689.1 | 9 | 1682 | |||
XM_011530883.1 | XP_011529185.1 | 9 | 1682 | |||
XM_017029322.1 | XP_016884811.1 | 3 | 625 | |||
XM_017029323.1 | XP_016884812.1 | 3 | 625 | |||
XM_017029324.1 | XP_016884813.1 | 3 | 645 | |||
GenBank mRNA | AB033058.1 | - | 3 | 634 | ||
AB209328.1 | - | 9 | 1677 | |||
AK304020.1 | - | 3 | 349 | |||
AK315041.1 | - | 9 | 1616 | |||
AK316518.1 | - | 3 | 335 | |||
BC093864.1 | - | 9 | 1378 | 7939709 | ||
BC093866.1 | - | 9 | 1378 | 7939711 | ||
CN357435.1 | - | 3 | 631 | |||
U49089.1 | - | 9 | 1563 |