Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNB SIRNA Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
UniGene ID:
Hs.307720
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146771
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256303.1
NP_001243232.1
12
1477
NM_001256304.1
NP_001243233.1
11
1214
NM_001256308.2
NP_001243237.1
11
1348
NM_001320932.1
NP_001307861.1
11
1322
NM_001320933.1
NP_001307862.1
11
1387
NM_001320934.1
NP_001307863.1
11
1387
NM_001320935.1
NP_001307864.1
10
1236
NM_001320936.1
NP_001307865.1
12
1477
NM_021907.4
NP_068707.1
12
1477
NM_033147.3
NP_149159.2
12
1477
NM_033148.3
NP_149160.1
11
1387
NM_183360.2
NP_899204.1
12
1477
NM_183361.2
NP_899205.1
11
1387
XM_005264182.1
XP_005264239.1
12
1394
XM_011532671.1
XP_011530973.1
12
1394
XM_011532672.2
XP_011530974.1
12
1394
XM_011532673.1
XP_011530975.1
12
1394
XM_011532674.1
XP_011530976.1
12
1394
XM_011532675.1
XP_011530977.1
12
1394
XM_011532676.1
XP_011530978.1
11
1304
XM_011532677.2
XP_011530979.1
11
1850
XM_011532679.1
XP_011530981.1
12
1394
XM_011532680.2
XP_011530982.1
11
1315
XM_011532681.2
XP_011530983.1
11
1308
XM_017003529.1
XP_016859018.1
11
1304
XM_017003530.1
XP_016859019.1
11
1304
XM_017003531.1
XP_016859020.1
11
1304
XM_017003532.1
XP_016859021.1
11
1304
XM_017003533.1
XP_016859022.1
11
1304
XM_017003534.1
XP_016859023.1
11
1304
XM_017003535.1
XP_016859024.1
11
1304
XM_017003536.1
XP_016859025.1
11
1304
XM_017003537.1
XP_016859026.1
8
1747
XM_017003538.1
XP_016859027.1
11
1240
XM_017003539.1
XP_016859028.1
10
1161
XM_017003540.1
XP_016859029.1
10
1218
XM_017003541.1
XP_016859030.1
10
1202
XM_017003542.1
XP_016859031.1
10
1156
XM_017003543.1
XP_016859032.1
9
1050
XM_017003544.1
XP_016859033.1
10
1156
XM_017003545.1
XP_016859034.1
9
1050
GenBank mRNA
AF022728.1
-
12
1375
AF070567.1
-
6
619
AK094494.1
-
11
1195
AK225724.1
-
11
1225
AK294174.1
-
10
1141
AK294357.1
-
11
1158
AK300019.1
-
12
1354
AK301386.1
-
11
1350
BC016655.1
-
11
1280
3868207
BC049366.1
-
12
1468
4817552
BT009805.1
-
10
1124
BX389926.1
-
5
507
DQ160290.1
-
11
1214
R15062.1
-
3
237
Y12712.1
-
11
1307
Y15718.1
-
11
1214
Y15722.1
-
11
1214
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline