Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243645.1 | NP_001230574.1 | 4 | 780 | ||
NM_021914.7 | NP_068733.1 | 4 | 803 | |||
NM_138638.4 | NP_619579.1 | 4 | 945 | |||
NR_028130.1 | 4 | 802 | ||||
NR_028131.1 | 4 | 691 | ||||
XM_011536363.2 | XP_011534665.1 | 4 | 741 | |||
GenBank mRNA | AF087867.1 | - | 4 | 698 | ||
AF134802.1 | - | 4 | 814 | |||
AF134803.1 | - | 4 | 708 | |||
AL117457.1 | - | 1 | 28 | |||
BC011444.1 | - | 4 | 729 | 4245104 | ||
BC022364.1 | - | 4 | 812 | 4730117 | ||
BC022876.1 | - | 4 | 816 | 4109108 | ||
BC025683.1 | - | 4 | 691 | |||
CB992605.1 | - | 4 | 645 |