Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033399.1 | 18 | 1981 | |||
GenBank mRNA | BC065494.1 | - | 9 | 581 | ||
U33834.1 | - | 15 | 1342 |
Gene Symbol: LOC642846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024374.1 | 18 | 2025 |