Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001224.4 | NP_001215.1 | 6 | 678 | ||
NM_032982.3 | NP_116764.2 | 6 | 924 | |||
NM_032983.3 | NP_116765.2 | 5 | 706 | |||
GenBank mRNA | AB209640.1 | - | 1 | 106 | ||
AF314174.1 | - | 5 | 414 | |||
AF314175.1 | - | 4 | 372 | |||
AK291274.1 | - | 6 | 716 | |||
AK303391.1 | - | 6 | 587 | |||
BC002427.2 | - | 6 | 696 | 3346279 | ||
BT007240.1 | - | 6 | 632 | |||
CR541748.1 | - | 5 | 590 | |||
KU177957.1 | - | 6 | 683 | |||
KU177958.1 | - | 4 | 433 | |||
U13021.1 | - | 6 | 645 | |||
U13022.1 | - | 6 | 678 |