Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001224.4 | NP_001215.1 | 11, 12 | 1291 | ||
NM_032982.3 | NP_116764.2 | 10, 11 | 1476 | |||
NM_032983.3 | NP_116765.2 | 9, 10 | 1258 | |||
GenBank mRNA | AB209640.1 | - | 4, 5 | 4148 | ||
AF314174.1 | - | 9, 10 | 966 | |||
AF314175.1 | - | 8, 9 | 924 | |||
AK291274.1 | - | 10, 11 | 1268 | |||
AK303265.1 | - | 4, 5 | 540 | |||
AK303391.1 | - | 10, 11 | 1139 | |||
BC002427.2 | - | 10, 11 | 1248 | 3346279 | ||
BT007240.1 | - | 10, 11 | 1184 | |||
KU177957.1 | - | 10, 11 | 1235 | |||
KU177958.1 | - | 7, 8 | 875 | |||
U13021.1 | - | 10, 11 | 1197 | |||
U13022.1 | - | 11, 12 | 1291 |