Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018115.2 | NP_001018125.1 | 3 | 310 | ||
NM_001319984.1 | NP_001306913.1 | 3 | 382 | |||
NM_033084.4 | NP_149075.2 | 3 | 310 | |||
GenBank mRNA | AF230336.1 | - | 3 | 253 | ||
AF340183.1 | - | 3 | 253 | |||
AK307512.1 | - | 4 | 283 | |||
BC013582.2 | - | 3 | 241 | |||
BX956311.1 | - | 3 | 281 | |||
DA749163.1 | - | 4 | 507 | |||
DA864640.1 | - | 3 | 311 |