Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190807.2 | NP_001177736.1 | 1 | 242 | ||
NM_001914.3 | NP_001905.1 | 1 | 242 | |||
NM_148923.3 | NP_683725.1 | 1 | 242 | |||
XM_011525835.2 | XP_011524137.1 | 1 | 206 | |||
GenBank mRNA | AB209617.1 | - | 1 | 183 | ||
AK026310.1 | - | 1 | 166 | |||
BC015182.1 | - | 1 | 170 | 3912638 | ||
CA771478.1 | - | 1 | 208 | |||
CB112646.1 | - | 1 | 242 | |||
CR456990.1 | - | 1 | 101 | |||
M22865.1 | - | 1 | 153 | |||
M60174.1 | - | 1 | 101 |