Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1T1 SIRNA Assays ›
Gene Symbol:
RUNX1T1
Gene Name:
RUNX1 translocation partner 1
Gene Aliases:
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location:
Chr.8: 91954967 - 92103365 on Build GRCh38
UniGene ID:
Hs.368431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146845
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUNX1T1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198625.1
NP_001185554.1
7
1070
NM_001198626.1
NP_001185555.1
5
928
NM_001198627.1
NP_001185556.1
6
988
NM_001198628.1
NP_001185557.1
4
585
NM_001198629.1
NP_001185558.1
4
711
NM_001198630.1
NP_001185559.1
5
771
NM_001198631.1
NP_001185560.1
4
784
NM_001198632.1
NP_001185561.1
4
702
NM_001198633.1
NP_001185562.1
3
673
NM_001198634.1
NP_001185563.1
3
611
NM_001198679.1
NP_001185608.1
4
755
NM_004349.3
NP_004340.1
3
721
NM_175634.2
NP_783552.1
4
846
NM_175635.2
NP_783553.1
3
538
NM_175636.2
NP_783554.1
2
620
XM_006716676.3
XP_006716739.1
5
655
XM_011517351.2
XP_011515653.1
5
700
XM_011517352.2
XP_011515654.1
5
940
XM_011517353.2
XP_011515655.1
4
648
XM_017013930.1
XP_016869419.1
4
956
XM_017013931.1
XP_016869420.1
3
478
XM_017013932.1
XP_016869421.1
5
843
XM_017013933.1
XP_016869422.1
6
907
XM_017013934.1
XP_016869423.1
6
1015
XM_017013935.1
XP_016869424.1
5
820
XM_017013936.1
XP_016869425.1
4
914
XM_017013937.1
XP_016869426.1
5
740
XM_017013938.1
XP_016869427.1
4
604
GenBank mRNA
AK297616.1
-
3
435
AK312592.1
-
7
832
BC005850.1
-
3
500
2961112
BC067078.1
-
3
491
2964656
BT009871.1
-
2
280
CR456792.1
-
2
310
D14289.1
-
2
577
D14821.1
-
3
475
D43638.1
-
3
721
KF946505.1
-
3
339
KF946507.1
-
3
339
KF946508.1
-
3
339
KF946510.1
-
3
339
KF946511.1
-
3
325
KF946512.1
-
4
1629
KF946513.1
-
3
2009
KF946514.1
-
3
325
KF946515.1
-
2
565
KF946516.1
-
2
565
KF946517.1
-
2
565
KF946518.1
-
2
565
X79990.1
-
2
303
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
transcription, DNA-templated
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline