Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCKBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318029.1 | NP_001304958.1 | 4 | 1490 | ||
NM_176875.3 | NP_795344.1 | 5 | 1742 | |||
XM_005253210.1 | XP_005253267.1 | 4 | 2012 | |||
XM_017018516.1 | XP_016874005.1 | 5 | 2228 | |||
GenBank mRNA | AF239668.1 | - | 4 | 1697 | ||
AK226020.1 | - | 6 | 1504 | |||
AK292064.1 | - | 5 | 1560 | |||
BC000740.2 | - | 5 | 1543 | 3504160 | ||
CD013884.1 | - | 4 | 1358 | |||
D13305.1 | - | 5 | 1679 | |||
L04473.1 | - | 5 | 1486 |
Set Membership: |
G-protein coupled receptor |