Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005227.2 | NP_005218.1 | 4 | 1227 | ||
NM_182689.1 | NP_872631.1 | 4 | 1061 | |||
NM_182690.2 | NP_872632.2 | 4 | 1081 | |||
GenBank mRNA | AJ006352.1 | - | 4 | 1165 | ||
AJ006353.1 | - | 4 | 1020 | |||
BQ940212.1 | - | 3 | 451 |