Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000044.3 | NP_000035.2 | 8 | 3789 | ||
NM_001011645.2 | NP_001011645.1 | 8 | 1240 | |||
GenBank mRNA | AF162704.1 | - | 10 | 2635 | ||
BC132975.1 | - | 8 | 3119 | 40146997 | ||
GU208210.1 | - | 5 | 2258 | |||
HQ692823.1 | - | 8 | 1078 | |||
KF720403.1 | - | 5 | 700 | |||
KF720404.1 | - | 5 | 700 | |||
KF720405.1 | - | 5 | 700 | |||
KJ534771.1 | - | 8 | 2671 | |||
L29496.1 | - | 8 | 2116 | |||
M20132.1 | - | 8 | 3033 | |||
M20260.1 | - | 8 | 1270 | |||
M21748.1 | - | 8 | 2827 | |||
M23263.1 | - | 8 | 3199 | |||
M34233.1 | - | 10 | 2740 | |||
M73069.1 | - | 9 | 2719 |