Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000066.3 | NP_000057.2 | 6 | 1023 | ||
NM_001278543.1 | NP_001265472.1 | 7 | 1245 | |||
NM_001278544.1 | NP_001265473.1 | 7 | 1152 | |||
XM_017002235.1 | XP_016857724.1 | 7 | 1091 | |||
GenBank mRNA | AK292740.1 | - | 6 | 878 | ||
AK298425.1 | - | 7 | 1069 | |||
AK298444.1 | - | 7 | 1245 | |||
AK313382.1 | - | 6 | 895 | |||
AK316209.1 | - | 7 | 1242 | |||
BC020701.1 | - | 6 | 898 | |||
BC130575.1 | - | 6 | 856 | 40146606 | ||
M16973.1 | - | 6 | 858 | |||
X04393.1 | - | 5 | 693 |