Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000214.2 | NP_000205.1 | 4, 5 | 1218 | ||
XM_017027707.1 | XP_016883196.1 | 3, 4 | 1323 | |||
GenBank mRNA | AF003837.1 | - | 4, 5 | 1161 | ||
AF028593.1 | - | 4, 5 | 965 | |||
AK302554.1 | - | 2, 3 | 568 | |||
AK312722.1 | - | 4, 5 | 787 | |||
BC098393.1 | - | 4, 5 | 1121 | 30528888 | ||
BC126205.1 | - | 5, 6 | 726 | 8991921 | ||
BC126207.1 | - | 4, 5 | 954 | 8991923 | ||
U61276.1 | - | 4, 5 | 1072 | |||
U73936.1 | - | 4, 5 | 1115 | |||
U77720.1 | - | 4, 5 | 664 |