Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000214.2 | NP_000205.1 | 26 | 4955 | ||
XM_017027707.1 | XP_016883196.1 | 25 | 5060 | |||
GenBank mRNA | AF003837.1 | - | 26 | 4897 | ||
AF028593.1 | - | 26 | 4702 | |||
BC098393.1 | - | 26 | 4858 | 30528888 | ||
U73936.1 | - | 26 | 4851 | |||
U77720.1 | - | 26 | 4401 |