Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000476.2 | NP_000467.1 | 6 | 605 | ||
NM_001318121.1 | NP_001305050.1 | 6 | 592 | |||
NM_001318122.1 | NP_001305051.1 | 5 | 600 | |||
XM_017014427.1 | XP_016869916.1 | 6 | 2048 | |||
XM_017014428.1 | XP_016869917.1 | 7 | 952 | |||
GenBank mRNA | AB021871.1 | - | 4 | 452 | ||
AK223501.1 | - | 5 | 480 | |||
AK313758.1 | - | 6 | 536 | |||
BC001116.2 | - | 6 | 523 | 2988248 | ||
BC100022.1 | - | 6 | 552 | 30561924 | ||
BG772183.1 | - | 5 | 600 | |||
BT019580.1 | - | 4 | 452 | |||
CR541977.1 | - | 4 | 452 | |||
CR542004.1 | - | 4 | 452 | |||
CR975542.1 | - | 6 | 592 | |||
EU794628.1 | - | 6 | 605 |